Learn More
Meso Scale Discovery UPLEX Rat PYY TTL ASY384well
SDP
Catalog No. 5000078990
Encompass
The Meso Scale Discovery U-PLEX Rat PYY Total 384-Well Assay is an electrochemiluminescent immunoassay for measuring total Peptide YY (PYY) in rat serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a rapid sensitive and cost-effective measurement with high precision The detection range is 0 3 to 10 000 pg/mL The kit includes a 384-well microplate with capture antibodies detection antibodies with SULFO-TAG calibrators and diluent Total hands-on time is 2 5 hours
Catalog No. 50-000-78990 Supplier Meso Scale Discovery Supplier No. K2536BK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat PYY Total 384-Well Assay is an electrochemiluminescent immunoassay for measuring total Peptide YY (PYY) in rat serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a rapid sensitive and cost-effective measurement with high precision The detection range is 0 3 to 10 000 pg/mL The kit includes a 384-well microplate with capture antibodies detection antibodies with SULFO-TAG calibrators and diluent Total hands-on time is 2 5 hours