Learn More
Meso Scale Discovery UPLEX Rat PYY TTL ASY384well
SDP
Catalog No. 5000081157
Encompass
The Meso Scale Discovery U-PLEX Rat PYY Total 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in rat serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 1 5 pg/mL and includes a 384-well plate coated with capture antibody detection antibody Read Buffer and Calibrator Diluent This product allows for high-throughput analysis of total PYY levels in rat biological samples
Catalog No. 50-000-81157 Supplier Meso Scale Discovery Supplier No. K2536BK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat PYY Total 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in rat serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 1 5 pg/mL and includes a 384-well plate coated with capture antibody detection antibody Read Buffer and Calibrator Diluent This product allows for high-throughput analysis of total PYY levels in rat biological samples