Learn More
Meso Scale Discovery UPLEX Rat PYY TTL ASYQP
SDP
Catalog No. 5000081503
Encompass
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is a multiplex immunoassay that measures total PYY levels in rat serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This rat PYY assay allows for the quantitative measurement of total PYY levels with high precision and accuracy The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the immunoassay
Catalog No. 50-000-81503 Supplier Meso Scale Discovery Supplier No. K1536BK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is a multiplex immunoassay that measures total PYY levels in rat serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This rat PYY assay allows for the quantitative measurement of total PYY levels with high precision and accuracy The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the immunoassay