Learn More
Meso Scale Discovery UPLEX Target Cell Checkpoint
SDP
Catalog No. 5000082217
Encompass
The Mesoscale K15340K-1 is a high-performance thermoelectric cooler with a 40W heat pumping capacity It features a thermoelectric module with a cold side heat sink and fan for efficient heat dissipation The module has 127 thermocouples made of Bismuth Telluride between ceramic substrates achieving a 70 C temperature differential Measuring 40 x 40 x 3 4mm it s ideal for cooling small spaces in scientific medical consumer and electronic applications It operates on a 12VDC power supply with control via DC current adjustment
Catalog No. 50-000-82217 Supplier Meso Scale Discovery Supplier No. K15340K1
The Mesoscale K15340K-1 is a high-performance thermoelectric cooler with a 40W heat pumping capacity It features a thermoelectric module with a cold side heat sink and fan for efficient heat dissipation The module has 127 thermocouples made of Bismuth Telluride between ceramic substrates achieving a 70 C temperature differential Measuring 40 x 40 x 3 4mm it s ideal for cooling small spaces in scientific medical consumer and electronic applications It operates on a 12VDC power supply with control via DC current adjustment