Learn More
Meso Scale Discovery UPLEX TCell Combo hu
SDP
Catalog No. 5000082282
Encompass
The U-PLEX T Cell Combo Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human IFN-gamma IL-2 IL-4 IL-5 IL-10 IL-13 IL-17A TNF-alpha and GM-CSF biomarkers from cell culture supernatants serum plasma and other biological samples The electrochemiluminescent assay utilizes patterned arrays and MSD s proprietary SULFO-TAG labels to provide sensitive and reproducible results from a small sample volume This 9-plex kit includes a 96-well plate calibrators controls 10X read buffer 10X wash buffer and reagents for 500 tests It is for research use only
Catalog No. 50-000-82282 Supplier Meso Scale Discovery Supplier No. K15093K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX T Cell Combo Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human IFN-gamma IL-2 IL-4 IL-5 IL-10 IL-13 IL-17A TNF-alpha and GM-CSF biomarkers from cell culture supernatants serum plasma and other biological samples The electrochemiluminescent assay utilizes patterned arrays and MSD s proprietary SULFO-TAG labels to provide sensitive and reproducible results from a small sample volume This 9-plex kit includes a 96-well plate calibrators controls 10X read buffer 10X wash buffer and reagents for 500 tests It is for research use only