Learn More
Meso Scale Discovery UPLEX TCell Combo ms
SDP
Catalog No. 5000081826
Encompass
The U-PLEX T Cell Combo Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous detection of mouse IFN-gamma IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IL-17A IL-17F IL-21 IL-22 IL-23 IL-27 IL-31 and TNF-alpha in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls The electrochemiluminescent assay allows sensitive and accurate quantification of T cell cytokines in less than 5 hours Recommended for examining T cell responses in immunology and inflammation research
Catalog No. 50-000-81826 Supplier Meso Scale Discovery Supplier No. K15098K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX T Cell Combo Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous detection of mouse IFN-gamma IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IL-17A IL-17F IL-21 IL-22 IL-23 IL-27 IL-31 and TNF-alpha in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls The electrochemiluminescent assay allows sensitive and accurate quantification of T cell cytokines in less than 5 hours Recommended for examining T cell responses in immunology and inflammation research