Learn More
Meso Scale Discovery UPLEX TCell Combo NHP
SDP
Catalog No. 5000078318
Encompass
The U-PLEX T Cell Combo NHP Kit from Meso Scale Discovery is a multiplex assay kit for measuring monkey interferon gamma interleukins 2 4 5 10 13 17A and tumor necrosis factor alpha in cell culture supernatants serum plasma and other samples It uses MSD s SECTOR Imager for high sensitivity and broad range with low sample volume The kit includes a 96-well plate capture and detection antibodies buffers diluent calibrators and controls enabling efficient analysis of T cell immune response in non-human primate samples
Catalog No. 50-000-78318 Supplier Meso Scale Discovery Supplier No. K15095K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX T Cell Combo NHP Kit from Meso Scale Discovery is a multiplex assay kit for measuring monkey interferon gamma interleukins 2 4 5 10 13 17A and tumor necrosis factor alpha in cell culture supernatants serum plasma and other samples It uses MSD s SECTOR Imager for high sensitivity and broad range with low sample volume The kit includes a 96-well plate capture and detection antibodies buffers diluent calibrators and controls enabling efficient analysis of T cell immune response in non-human primate samples