Learn More
Meso Scale Discovery UPLEX TGFB Combo hu 25 PL
SDP
Catalog No. 5000081403
Encompass
The Meso Scale Discovery U-PLEX TGF-beta Combo Human Kit is a multiplex immunoassay measuring active and total TGF-beta 1 and pro and active MMP-9 in various samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible quantification from a small sample With a broad dynamic range it s ideal for research in cancer immunology and fibrosis The kit includes a 96-well plate calibrators detection antibodies read buffer sample diluent wash buffer and plate seals for one 96-well plate
Catalog No. 50-000-81403 Supplier Meso Scale Discovery Supplier No. K15241K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX TGF-beta Combo Human Kit is a multiplex immunoassay measuring active and total TGF-beta 1 and pro and active MMP-9 in various samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible quantification from a small sample With a broad dynamic range it s ideal for research in cancer immunology and fibrosis The kit includes a 96-well plate calibrators detection antibodies read buffer sample diluent wash buffer and plate seals for one 96-well plate