Learn More
Meso Scale Discovery UPLEX TGFB Combo NHP
SDP
Catalog No. 5000082354
Encompass
The Meso Scale Discovery U-PLEX TGF-beta Combo NHP Assay is a multiplex immunoassay kit for measuring active and total TGF-beta 1 2 and 3 in non-human primate serum plasma and cell culture supernatants It uses MSD s SECTOR Imager for high sensitivity broad dynamic range and low sample volume The assay quantifies TGF-beta isoforms crucial cytokines in biological processes The kit includes a plate with capture antibodies detection antibodies calibrators controls 10X read buffer sample diluent and wash buffer
Catalog No. 50-000-82354 Supplier Meso Scale Discovery Supplier No. K15243K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX TGF-beta Combo NHP Assay is a multiplex immunoassay kit for measuring active and total TGF-beta 1 2 and 3 in non-human primate serum plasma and cell culture supernatants It uses MSD s SECTOR Imager for high sensitivity broad dynamic range and low sample volume The assay quantifies TGF-beta isoforms crucial cytokines in biological processes The kit includes a plate with capture antibodies detection antibodies calibrators controls 10X read buffer sample diluent and wash buffer