Learn More
Meso Scale Discovery UPLEX TH1/TH2 Combo hu
SDP
Catalog No. 5000078770
Encompass
The U-PLEX TH1/TH2 Combo Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantification of human IFN- IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 and TNF- This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls It allows efficient profiling of Th1/Th2 immune responses from cell culture supernatants serum plasma and other biological samples
Catalog No. 50-000-78770 Supplier Meso Scale Discovery Supplier No. K15010K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH1/TH2 Combo Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantification of human IFN- IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 and TNF- This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls It allows efficient profiling of Th1/Th2 immune responses from cell culture supernatants serum plasma and other biological samples