Learn More
Meso Scale Discovery UPLEX TH1/TH2 Combo ms
SDP
Catalog No. 5000079231
Encompass
The U-PLEX TH1/TH2 Combo Mouse Assay is a multiplex immunoassay kit for quantifying 7 mouse cytokines related to T helper cell responses including IFN- IL-2 IL-4 IL-5 IL-10 IL-12p70 and TNF- It uses electrochemiluminescence detection and is compatible with Meso Scale Discovery instruments The assay requires 50 L of sample per well and can be completed in 3 5 hours It includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrator solutions for standard curves allowing efficient analysis from a small sample
Catalog No. 50-000-79231 Supplier Meso Scale Discovery Supplier No. K15071K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH1/TH2 Combo Mouse Assay is a multiplex immunoassay kit for quantifying 7 mouse cytokines related to T helper cell responses including IFN- IL-2 IL-4 IL-5 IL-10 IL-12p70 and TNF- It uses electrochemiluminescence detection and is compatible with Meso Scale Discovery instruments The assay requires 50 L of sample per well and can be completed in 3 5 hours It includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrator solutions for standard curves allowing efficient analysis from a small sample