Learn More
Meso Scale Discovery UPLEX TH17 Combo 1 hu
SDP
Catalog No. 5000078259
Encompass
The U-PLEX TH17 Combo 1 Human Kit from Meso Scale Discovery is a multiplex assay kit that enables simultaneous measurement of IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 IL-33 sCD40L and TNF-alpha from cell culture supernatants serum plasma (EDTA heparin) and saliva This electrochemiluminescence (ECL) based immunoassay utilizes MSD s patented SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrators and controls
Catalog No. 50-000-78259 Supplier Meso Scale Discovery Supplier No. K15075K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH17 Combo 1 Human Kit from Meso Scale Discovery is a multiplex assay kit that enables simultaneous measurement of IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 IL-33 sCD40L and TNF-alpha from cell culture supernatants serum plasma (EDTA heparin) and saliva This electrochemiluminescence (ECL) based immunoassay utilizes MSD s patented SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrators and controls