Learn More
Meso Scale Discovery UPLEX TH17 Combo 1 ms
SDP
Catalog No. 5000081699
Encompass
The U-PLEX TH17 Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of IL-17A IL-17F IL-21 IL-22 IL-23 and IL-31 in mouse samples The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay allows sensitive precise and accurate quantification of the 6 mouse Th17-related cytokines from a single small-volume sample in under 5 hours The kit components are optimized for use with MSD s QuickPlex SQ 120 Instrument and MSD Discovery Workbench Software
Catalog No. 50-000-81699 Supplier Meso Scale Discovery Supplier No. K15077K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH17 Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of IL-17A IL-17F IL-21 IL-22 IL-23 and IL-31 in mouse samples The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay allows sensitive precise and accurate quantification of the 6 mouse Th17-related cytokines from a single small-volume sample in under 5 hours The kit components are optimized for use with MSD s QuickPlex SQ 120 Instrument and MSD Discovery Workbench Software