Learn More
Meso Scale Discovery UPLEX TH17 Combo 1 NHP
SDP
Catalog No. 5000078281
Encompass
The U-PLEX TH17 Combo 1 NHP is a multiplex assay kit by Meso Scale Diagnostics that enables simultaneous quantitative measurement of IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 IL-33 sCD40L and TNF-alpha from cell culture supernatants serum plasma (EDTA heparin) and saliva It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer It is suitable for use in biomarker discovery and immunology research
Catalog No. 50-000-78281 Supplier Meso Scale Discovery Supplier No. K15079K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH17 Combo 1 NHP is a multiplex assay kit by Meso Scale Diagnostics that enables simultaneous quantitative measurement of IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 IL-33 sCD40L and TNF-alpha from cell culture supernatants serum plasma (EDTA heparin) and saliva It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer It is suitable for use in biomarker discovery and immunology research