Learn More
Meso Scale Discovery UPLEX TH17 Combo 2 hu
SDP
Catalog No. 5000078254
Encompass
The U-PLEX TH17 Combo 2 Human kit from Meso Scale Discovery is a multiplex immunoassay that measures the levels of IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 IL-33 and TNF-alpha in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay allows sensitive accurate and rapid quantification of the 8 cytokines related to T helper 17 cell immune responses The kit has a broad dynamic range of 0 01-2500 pg/mL and requires only 25 L sample size per well
Catalog No. 50-000-78254 Supplier Meso Scale Discovery Supplier No. K15076K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH17 Combo 2 Human kit from Meso Scale Discovery is a multiplex immunoassay that measures the levels of IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 IL-33 and TNF-alpha in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay allows sensitive accurate and rapid quantification of the 8 cytokines related to T helper 17 cell immune responses The kit has a broad dynamic range of 0 01-2500 pg/mL and requires only 25 L sample size per well