Learn More
Meso Scale Discovery UPLEX Vascular Injury
SDP
Catalog No. 5000081168
Encompass
The Meso Scale Discovery U-PLEX Vascular Injury Combo 1 Human Kit is a multiplex immunoassay for measuring biomarkers related to vascular injury and endothelial dysfunction It quantifies VCAM-1 ICAM-1 E-Selectin P-Selectin and Thrombomodulin from serum plasma and cell culture supernatants in one well The electrochemiluminescent detection offers high sensitivity and a broad range This kit aids in studying cardiovascular disease diabetes sepsis and other vascular injury conditions It includes a 96-well plate calibrators controls detection antibodies buffers and seals
Catalog No. 50-000-81168 Supplier Meso Scale Discovery Supplier No. K15390K1
The Meso Scale Discovery U-PLEX Vascular Injury Combo 1 Human Kit is a multiplex immunoassay for measuring biomarkers related to vascular injury and endothelial dysfunction It quantifies VCAM-1 ICAM-1 E-Selectin P-Selectin and Thrombomodulin from serum plasma and cell culture supernatants in one well The electrochemiluminescent detection offers high sensitivity and a broad range This kit aids in studying cardiovascular disease diabetes sepsis and other vascular injury conditions It includes a 96-well plate calibrators controls detection antibodies buffers and seals