Learn More
Meso Scale Discovery UPLEX Vascular Injury
SDP
Catalog No. 5000081558
Encompass
The Meso Scale Discovery U-PLEX VCAM-1 Assay Kit is a multiplex immunoassay that measures VCAM-1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes approximately 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls The kit has high specificity and no cross-reactivity It allows simultaneous detection of multiple biomarkers
Catalog No. 50-000-81558 Supplier Meso Scale Discovery Supplier No. K15390K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX VCAM-1 Assay Kit is a multiplex immunoassay that measures VCAM-1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes approximately 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls The kit has high specificity and no cross-reactivity It allows simultaneous detection of multiple biomarkers