Learn More
Meso Scale Discovery UPLEX Vascular Injury
SDP
Catalog No. 5000081559
Encompass
The Meso Scale Discovery U-PLEX Vascular Injury Combo 1 Human Kit is a multiplex immunoassay that measures biomarkers related to vascular injury and inflammation This kit allows for the simultaneous quantitative measurement of CRP ICAM-1 VCAM-1 and SAA from serum plasma and cell culture supernatants in a single well The electrochemiluminescent detection method provides high sensitivity and a large dynamic range This product requires the MSD QuickPlex SQ120 instrument and provides sufficient reagents for one 96-well plate With a short 3 5 hour protocol this kit enables efficient quantification of multiple biomarkers related to vascular inflammation and injury
Catalog No. 50-000-81559 Supplier Meso Scale Discovery Supplier No. K15390K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Vascular Injury Combo 1 Human Kit is a multiplex immunoassay that measures biomarkers related to vascular injury and inflammation This kit allows for the simultaneous quantitative measurement of CRP ICAM-1 VCAM-1 and SAA from serum plasma and cell culture supernatants in a single well The electrochemiluminescent detection method provides high sensitivity and a large dynamic range This product requires the MSD QuickPlex SQ120 instrument and provides sufficient reagents for one 96-well plate With a short 3 5 hour protocol this kit enables efficient quantification of multiple biomarkers related to vascular inflammation and injury