Learn More
Waters Corp µPorasil Silica Radial-Pak Column, 125Å, 10 µm, 25 mm X 100 mm, 1/pk
Catalog No. 50785453 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters µPorasil 10µm silica packing material provides high-efficiency and low backpressure.
Catalog No. 50-785-453 Supplier Waters Corp Supplier No. WAT038504
Waters µPorasil 10µm silica packing material provides high-efficiency and low backpressure.