Learn More
Waters Corp Waters μPorasil Guard Column, 10 μm, 125 Å, 3.9 × 20 mm
Catalog No. 50785629 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The μPorasil silica packing material provides high-efficiency and low backpressure, making it suitable for various chromatographic applications.
  • Silica packing material.
  • 10 μm particle size for efficient separations.
  • Low backpressure for system protection.
  • 125 Å pore size.
  • 3.9 × 20 mm column dimensions.
  • Normal-phase separation mode.
  • pH range from 2 to 8.
  • Maximum pressure of 6000 psi (415 bar).
  • Compatible with HPLC systems.
  • Supplied as a 2-pack.
Catalog No. 50-785-629 Supplier Waters Corp Supplier No. WAT046860
The μPorasil silica packing material provides high-efficiency and low backpressure, making it suitable for various chromatographic applications.
  • Silica packing material.
  • 10 μm particle size for efficient separations.
  • Low backpressure for system protection.
  • 125 Å pore size.
  • 3.9 × 20 mm column dimensions.
  • Normal-phase separation mode.
  • pH range from 2 to 8.
  • Maximum pressure of 6000 psi (415 bar).
  • Compatible with HPLC systems.
  • Supplied as a 2-pack.