Learn More
Aviva Systems Biology Corp UPP1 Antibody - middle region (ARP42126_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42126_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UPP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UPP1. Alternate names: UDRPASE, UP, UPASE, UPPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58118