Learn More
Perkin Elmer US LLC Upper And Lower Anvil Set
Catalog No. 502113505
Encompass_Preferred
The Evacuable Potassium Bromide Die prepares KBr pellets of the hightest quality and clarity. A pellet, 13 mm in diameter, is formed inside its evacuated chamber and pressed between a highly polished anvil and plunger. This long-lasting die is precisely constructed to produce pellets of uniform quality.
Catalog No. 50-211-3505 Supplier Perkin Elmer US LLC Supplier No. 04820519
The Evacuable Potassium Bromide Die prepares KBr pellets of the hightest quality and clarity. A pellet, 13 mm in diameter, is formed inside its evacuated chamber and pressed between a highly polished anvil and plunger. This long-lasting die is precisely constructed to produce pellets of uniform quality.