Learn More
Waters Corp Waters Upper Needle Seal Support Sleeve & Cup, UPLC System, Sample Manager Component
Catalog No. NC1663388 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a replacement upper needle seal support sleeve and overflow cup assembly. It is an integral component of the Sample Manager - FTN injection and wash mechanism designed for use with all current UPLC FTN Sample Managers.
  • Part of the sample manager - FTN injection and wash mechanism
  • Designed for UPLC FTN Sample Managers
  • Suitable for UPLC H-Class System, UPLC I-Class System, and Premier systems
  • Includes one unit per package
  • Replaced as required
Catalog No. NC1663388 Supplier Waters Corp Supplier No. 700005176
This product is a replacement upper needle seal support sleeve and overflow cup assembly. It is an integral component of the Sample Manager - FTN injection and wash mechanism designed for use with all current UPLC FTN Sample Managers.
  • Part of the sample manager - FTN injection and wash mechanism
  • Designed for UPLC FTN Sample Managers
  • Suitable for UPLC H-Class System, UPLC I-Class System, and Premier systems
  • Includes one unit per package
  • Replaced as required