Learn More
Labrepco Inc Upright Freezer Drawer Rack, Corrosion-Resistant Stainless Steel, Sliding Drawers, 12 Box Capacity, 35.2 Qty Purchased
SDP
Catalog No. NC1788340
Encompass_Preferred
The UPR FRZ DRW RACK 3.75IN BX is designed to enhance organization and accessibility in freezer storage for laboratory samples. Constructed from corrosion-resistant stainless steel, it accommodates standard 3.75 in high boxes in a 4x3 configuration, providing a capacity for 12 boxes. Its sliding drawers feature easy-grip handles and card label holders for efficient inventory management, ensuring a user-friendly experience while maximizing storage space.
Catalog No. NC1788340 Supplier Labrepco Inc Supplier No. UFD43375
The UPR FRZ DRW RACK 3.75IN BX is designed to enhance organization and accessibility in freezer storage for laboratory samples. Constructed from corrosion-resistant stainless steel, it accommodates standard 3.75 in high boxes in a 4x3 configuration, providing a capacity for 12 boxes. Its sliding drawers feature easy-grip handles and card label holders for efficient inventory management, ensuring a user-friendly experience while maximizing storage space.