Learn More
Research Products International Corp Storage Rack for Upright Freezers, Holds 96 Well or 384 Well Plates, Rack Dimensions (Inches) 22 x 5 1/2 x 11H
SDP
Catalog No. 50998162
Encompass
Storage Racks are constructed of corrsion resistant stainless steel. Designed to help maximize freezer space, these racks are for 96-Well or 384-Well plates.
Fits micro-plates with dimensions of 5 x 3 3/8 x 9/16 (inches).
Catalog No. 50-998-162 Supplier Research Products International Corp Supplier No. 247553
Storage Racks are constructed of corrsion resistant stainless steel. Designed to help maximize freezer space, these racks are for 96-Well or 384-Well plates.
Fits micro-plates with dimensions of 5 x 3 3/8 x 9/16 (inches).