Learn More
Fisher Science Education™ Upright Compound Microscope Achromat Objective
Shop All Fisherbrand ProductsDescription
- RMS
- DIN standard
- Available in 4X or 40X
- Achromat Microscope Objective
- Correction for color and spherical aberration
Specifications
Specifications
| Type | Achromat Objective |
| Autoclavable | Not Autoclavable |
| Length (Metric) | 160 mm |
| Magnification Power | 4X |
| Sterility | Non-sterile |
| Voltage | 0/1 V |
| Warranty | 5 years optical and mechanical, 1 year electrical from date of purchase |