Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Drawer Freezer Rack, Holds (12) 100-Cell Hinged Top Boxes, Rack Dims 23 1/16L x 9 3/4H x 6 7/16W" (585 x 247 x 164mm), with Compartment Dims 5 5/8L x 2 3/16H x 6 1/8W" (143 x 56 x 156mm) 4x3
Catalog No. NC2208106
Encompass_Preferred
Rack Designed to hold (12) 100 Cell Hinged Top Plastic Boxes (2 "H x 6W x 5 1/2L" (54 x 152 x 141mm), made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208106 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFDHT43
Rack Designed to hold (12) 100 Cell Hinged Top Plastic Boxes (2 "H x 6W x 5 1/2L" (54 x 152 x 141mm), made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.