Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Freezer Rack, Holds (30) 50-Cell Hinged Top Boxes, Rack Dims 21 7/16L x 12 5/8H x 5 7/8W" (545 x 320 x 150mm), Compartment Dims 3 1/2L x 2 5/16H x 5 9/16W" (89 x 58 x 142mm) 6x5
Catalog No. NC2208105
Encompass_Preferred
Rack Designed to hold (30) 50 Cell Hinged Top Plastic Boxes (2 1/5"H x5 9/16W x3 3/8L" (56 x 141 x 86mm), made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208105 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFDH5065
Rack Designed to hold (30) 50 Cell Hinged Top Plastic Boxes (2 1/5"H x5 9/16W x3 3/8L" (56 x 141 x 86mm), made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.