Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Drawer Freezer Rack(7) Drawers, Holds (84) Plates w/Lids, (112) no Lids, Rack Dims 25 1/4L x 9 3/4H x 5 7/16W"(642 x 247 x 138m) Compartment Dims 3 1/2L x 2 3/16H x 5 1/8W"(89 x 56 x 130mm)7x4
Catalog No. NC2208102
Encompass_Preferred
Rack Designed to hold 96 Well and 384 Well Plates (84) Plates with lids/(112) Plates no lids, or (28) 96 Well Microtube Boxes (2 1/8" H,) made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208102 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFDMP74
Rack Designed to hold 96 Well and 384 Well Plates (84) Plates with lids/(112) Plates no lids, or (28) 96 Well Microtube Boxes (2 1/8" H,) made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.