Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Drawer Freezer Rack, Holds (28) Plates 14.4-44.9mmH, Rack Dims 25 1/4L x 8 1/16H x5 7/16W" (642 x 205 x 138mm), Compartment Dims 3 1/2L x 1 3/4H x 5 1/8W" (90 x 45 x 130mm) 7x4
Catalog No. NC2208108
Encompass_Preferred
Rack Designed to hold (28) Matrix, Micronic, Fluidx Racks (9/16-1 3/4" H x 5 1/32L x 3 3/8W", (14.4-44.9 x 127.7 x 85.5mm) made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208108 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFDDP74
Rack Designed to hold (28) Matrix, Micronic, Fluidx Racks (9/16-1 3/4" H x 5 1/32L x 3 3/8W", (14.4-44.9 x 127.7 x 85.5mm) made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.