Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Stainless Steel Freezer Rack, (2) Drawers, Holds (120) 15ml Centrifuge Tubes, Rack Dimensions 16 11/16L x 9 15/16H x 5 1/2W" (424 x 252 x 140mm)
Catalog No. NC2208051
Encompass_Preferred
Rack Designed to hold (120) 15ml Centrifuge Tubes (60) Tubes in each of 2 Drawers, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208051 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFDT152
Rack Designed to hold (120) 15ml Centrifuge Tubes (60) Tubes in each of 2 Drawers, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.