Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Stainless Steel Freezer Rack, (2) Drawers Holds (78) 50ml Centrifuge Tubes, Rack Dimensions 22/316L x 9 15/16H x 5 1/2W" (564 x 252 x 140mm)
Catalog No. NC2208065
Encompass_Preferred
Rack Designed to hold (78) 50ml Centrifuge Tubes, (39) Tubes in each of 2 Drawers, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208065 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFDLT502
Rack Designed to hold (78) 50ml Centrifuge Tubes, (39) Tubes in each of 2 Drawers, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.