Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Stainless Steel Freezer Rack, (1) Drawer, Holds (51) 50ml Centrifuge Tubes, Rack Dimensions 26 15/16L x 4 15/16H x 5 1/2W" (685 x 126 x 140mm)
Catalog No. NC2208061
Encompass_Preferred
Rack Designed to hold (51) 50ml Centrifuge Tubes in 1 Drawer, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208061 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFDXLT501
Rack Designed to hold (51) 50ml Centrifuge Tubes in 1 Drawer, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.