Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Freezer Rack, (2) Drawers for Storage Bottles w/H <4 13/16" (122mm), Rack Dimensions 22 3/16L x 9 15/16H x 5 1/2W" (564 x 252 x 140mm), Drawer Area 20 3/4 x 4 1/2" (523 x 114mm) per Drawer
Catalog No. NC2208088
Encompass_Preferred
Rack Designed to store bottles < 4 13/16" (122mm)H, in each of 2 Drawers, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack. Great for organizing -20⁰, -30⁰ , -86⁰ Freezers and Cold Rooms
This item is not returnable. View return policy
Catalog No. NC2208088 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFDLB2
Rack Designed to store bottles < 4 13/16" (122mm)H, in each of 2 Drawers, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack. Great for organizing -20⁰, -30⁰ , -86⁰ Freezers and Cold Rooms