Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Drawer Style Freezer Rack, Holds (8) Large 3.75" Boxes, Rack Dims 24L x 8 3/8H x 6 1/8W" (609 x 213 x 155m) Compartment Dims 5 7/8L x 3 15/16H x 5 13/16W" (149 x 100 x 148mm) 4x2
Catalog No. NC2208087
Encompass_Preferred
Rack Designed to hold (8) Large 3.75H"Cardboard Boxes ((3 7/8H x 5 13/16L x 5 13/16W", (98 x 146 x 146mm)) made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.
This item is not returnable. View return policy
Catalog No. NC2208087 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFD42L375
Rack Designed to hold (8) Large 3.75H"Cardboard Boxes ((3 7/8H x 5 13/16L x 5 13/16W", (98 x 146 x 146mm)) made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.