Learn More
Labrepco Inc UFD-462-S Upright Freezer Rack, Stainless Steel, Fits 2" Cryo Boxes, 24-Box Capacity, Drawer Style
SDP
Catalog No. NC2306315
Encompass_Preferred
The UFD-462-S upright freezer drawer rack enhances laboratory efficiency by providing organized storage for 24 cryo boxes, specifically designed for 2 in high boxes. Constructed from durable stainless steel, it optimizes space in ultra-low temperature freezers, featuring convenient individual drawer access and integrated handles for easy retrieval. This product is especially compatible with PHCbi (Panasonic/Sanyo) VIP upright freezers, ensuring superior functionality for scientific storage needs.
Catalog No. NC2306315 Supplier Labrepco Inc Supplier No. UFD462S
The UFD-462-S upright freezer drawer rack enhances laboratory efficiency by providing organized storage for 24 cryo boxes, specifically designed for 2 in high boxes. Constructed from durable stainless steel, it optimizes space in ultra-low temperature freezers, featuring convenient individual drawer access and integrated handles for easy retrieval. This product is especially compatible with PHCbi (Panasonic/Sanyo) VIP upright freezers, ensuring superior functionality for scientific storage needs.