Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Combination Drawer Racks for 2″ Fiberboard Boxes & 15mL Centrifuge Tubes (2 x 2)
SDP
Catalog No. 503028397
Encompass_Preferred
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube Upright Combination Drawer Freezer Racks conveniently stores 2” Fiberboard boxes and 15ml centrifuge tubes. These high-quality racks are constructed of corrosion resistant stainless steel and include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. This freezer rack does not come standard with boxes or tubes. Dimensions L x H x W:  11 5/16 x 9 11/16 x 5 1/2 inch ( 287 x 246 x 140mm)Compartment Dimension L x H x W: 5 3/8  x 2 1/8 x 5 1/4 inch (137 x 54.4 x 133 mm)
Catalog No. 50-302-8397 Supplier Alkali Scientific Supplier No. UFDRT15222
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube Upright Combination Drawer Freezer Racks conveniently stores 2” Fiberboard boxes and 15ml centrifuge tubes. These high-quality racks are constructed of corrosion resistant stainless steel and include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. This freezer rack does not come standard with boxes or tubes. Dimensions L x H x W:  11 5/16 x 9 11/16 x 5 1/2 inch ( 287 x 246 x 140mm)Compartment Dimension L x H x W: 5 3/8  x 2 1/8 x 5 1/4 inch (137 x 54.4 x 133 mm)