Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Combination Drawer Racks for 2″ Fiberboard Boxes & 50mL Centrifuge Tubes (3 x 2)
SDP
Catalog No. 503029051
Encompass_Preferred
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube Upright Combination Drawer Freezer Racks conveniently stores 2” Fiberboard boxes and 50ml centrifuge tubes. These high-quality racks are constructed of corrosion resistant stainless steel and include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. This freezer rack does not come standard with boxes or tubes. Dimensions L x H x W : 16 11/16 x 9 11/16 x 5 1/2 inch (424 x 246 x 140mm)Compartment Size: L x H x W : 5 3/8 x 2 1/8 x 5 1/4 (137 x 54.4 x 133mm)
Catalog No. 50-302-9051 Supplier Alkali Scientific Supplier No. UFDRT50322
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube Upright Combination Drawer Freezer Racks conveniently stores 2” Fiberboard boxes and 50ml centrifuge tubes. These high-quality racks are constructed of corrosion resistant stainless steel and include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. This freezer rack does not come standard with boxes or tubes. Dimensions L x H x W : 16 11/16 x 9 11/16 x 5 1/2 inch (424 x 246 x 140mm)Compartment Size: L x H x W : 5 3/8 x 2 1/8 x 5 1/4 (137 x 54.4 x 133mm)