Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Combination Drawer Racks for 2″ Fiberboard Boxes and Storage Bottles – Rack Only 4 x 2
SDP
Catalog No. 503027917
Encompass_Preferred
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube The UFDR-LB-422 Combination Drawer freezer Racks conveniently stores up to (8) 2 inch boxes and multiple storage bottles. These high-quality freezer racks are constructed of corrosion resistant stainless steel, include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Boxes & storage bottles are not included with purchase. Dimension LxHxW: 22 3/16 x 9 11/16 x 5 1/2 inch (564 x 246 x 140 mm)Compartment Size lxhxw: 5 3/8 x 2 1/8 x 5 1/4 inch (137 x 54.4 x 133 mm)
Catalog No. 50-302-7917 Supplier Alkali Scientific Supplier No. UFDRLB422
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube The UFDR-LB-422 Combination Drawer freezer Racks conveniently stores up to (8) 2 inch boxes and multiple storage bottles. These high-quality freezer racks are constructed of corrosion resistant stainless steel, include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Boxes & storage bottles are not included with purchase. Dimension LxHxW: 22 3/16 x 9 11/16 x 5 1/2 inch (564 x 246 x 140 mm)Compartment Size lxhxw: 5 3/8 x 2 1/8 x 5 1/4 inch (137 x 54.4 x 133 mm)