Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Combination Drawer Racks for 2″ Boxes and Storage Bottles, 3x2 configuration
SDP
Catalog No. 503028149
Encompass_Preferred
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube The UFDR-B-322 Combination Drawer freezer Racks conveniently stores up to (6) 2 inch boxes and multiple storage bottles. These high-quality freezer racks are constructed of corrosion resistant stainless steel, include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Boxes & storage bottles are not included with purchase. Dimension LxHxW: 16 11/16  x 9 11/ 16 x 5 1 /2 inch (424 x246 x 140mm)Compartment Size lxhxw: 5 3/8 x 2 1/8 x 5 1/4 inch (137 x 54.4 x 133mm)
Catalog No. 50-302-8149 Supplier Alkali Scientific Supplier No. UFDRB322
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube The UFDR-B-322 Combination Drawer freezer Racks conveniently stores up to (6) 2 inch boxes and multiple storage bottles. These high-quality freezer racks are constructed of corrosion resistant stainless steel, include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Boxes & storage bottles are not included with purchase. Dimension LxHxW: 16 11/16  x 9 11/ 16 x 5 1 /2 inch (424 x246 x 140mm)Compartment Size lxhxw: 5 3/8 x 2 1/8 x 5 1/4 inch (137 x 54.4 x 133mm)