Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Combination Drawer Racks for 2"Fiber Cardboard Boxes & 15ml Centrifuge Tubes, 4x2 Configuration
SDP
Catalog No. 503028043
Encompass_Preferred
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube The UFDR-LT15-422 Upright Combination Drawer Freezer Racks conveniently stores 2” Fiberboard boxes and 15ml centrifuge tubes. These high-quality racks are constructed of corrosion resistant stainless steel and include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. This freezer rack does not come standard with boxes or tubes. Dimensions L x H x W : 22 3/16 x 9 11/16 x 5 1/2inch (564 x 246 x 140mm)Compartment Size: L x H x W : 5 3/8 x 2 1/8 x 5 1/4inch (137 x 54.4 x 133 mm)
Catalog No. 50-302-8043 Supplier Alkali Scientific Supplier No. UFDRLT15422
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube The UFDR-LT15-422 Upright Combination Drawer Freezer Racks conveniently stores 2” Fiberboard boxes and 15ml centrifuge tubes. These high-quality racks are constructed of corrosion resistant stainless steel and include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. This freezer rack does not come standard with boxes or tubes. Dimensions L x H x W : 22 3/16 x 9 11/16 x 5 1/2inch (564 x 246 x 140mm)Compartment Size: L x H x W : 5 3/8 x 2 1/8 x 5 1/4inch (137 x 54.4 x 133 mm)