Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Combination Drawer Racks for 2″ Boxes and Storage Bottles, 2x2 configuration
SDP
Catalog No. 503028050
Encompass_Preferred
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube The UFDR-B-222 Combination Drawer freezer Racks conveniently stores up to (4) 2 inch boxes and multiple storage bottles. These high-quality freezer racks are constructed of corrosion resistant stainless steel, include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Boxes & storage bottles are not included with purchase. Dimension LxHxW:11 5/16 x 9 11/16 x 5 1/2 inch (287 x246 x 140 mm)Compartment Size lxhxw: 5 3/8 x 2 1/8 x 5 1 /4 inch (137 x 54.4 x 133mm)
Catalog No. 50-302-8050 Supplier Alkali Scientific Supplier No. UFDRB222
For 2 ” Boxes, 15ml & 50ml Centrifuge Tube The UFDR-B-222 Combination Drawer freezer Racks conveniently stores up to (4) 2 inch boxes and multiple storage bottles. These high-quality freezer racks are constructed of corrosion resistant stainless steel, include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Boxes & storage bottles are not included with purchase. Dimension LxHxW:11 5/16 x 9 11/16 x 5 1/2 inch (287 x246 x 140 mm)Compartment Size lxhxw: 5 3/8 x 2 1/8 x 5 1 /4 inch (137 x 54.4 x 133mm)