Learn More
Labrepco Inc UPRIGHT FREEZER COMBINATION DR
SDP
Catalog No. 509020122
Encompass_Preferred
Combination drawer rack, Upright freezer; Labrepco; Crystal; For 2 in. boxes, 15mL and 50mL Centrifuge tubes; Capacity: 8 Boxes plus 39 Tubes (50mL); L x H x D: 559 x 246 x 140mm
Catalog No. 50-902-0122 Supplier Labrepco Inc Supplier No. UFDLT50422
Combination drawer rack, Upright freezer; Labrepco; Crystal; For 2 in. boxes, 15mL and 50mL Centrifuge tubes; Capacity: 8 Boxes plus 39 Tubes (50mL); L x H x D: 559 x 246 x 140mm