Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for 15 mL Centrifuge Tubes; 2 Drawers, holds 160 tubes
Catalog No. NC1490413 Shop All Cole-Parmer Products
Encompass_Preferred
Height (in) 9 15/16 x Length (in) 22 x Width (in)5 1/2. This freezer drawer rack made of sturdy corrosion-resistant stainless steel construction for long-term use. has drawers that slide out for quick access and storage of sample tubes. It can easily store large quantities of 15 and 50 mL centrifuge tubes.
Catalog No. NC1490413 Supplier Cole-Parmer Supplier No. 0439827
Height (in) 9 15/16 x Length (in) 22 x Width (in)5 1/2. This freezer drawer rack made of sturdy corrosion-resistant stainless steel construction for long-term use. has drawers that slide out for quick access and storage of sample tubes. It can easily store large quantities of 15 and 50 mL centrifuge tubes.