Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 0.2mL Tube 96-Well PCR Boxes, 7x4 configuration
SDP
Catalog No. 503027913
Encompass_Preferred
For 0.2ml Tube 96-Well PCR BoxesThe UFDPCR-74 Upright Freezer Drawer Racks for 25-Place Slide Boxes. Constructed of corrosion resistant stainsteel. Dimensions L x H x W: 25 3/4  x 12 7/16  x 5 7/16 inch (654 x 316 x 138mm)Compartment Size l x h x w: 3 5/8 x 2 7/8 x 5 3/16 inch (92 x 73 x 131 mm)
Catalog No. 50-302-7913 Supplier Alkali Scientific Supplier No. UFDPCR74
For 0.2ml Tube 96-Well PCR BoxesThe UFDPCR-74 Upright Freezer Drawer Racks for 25-Place Slide Boxes. Constructed of corrosion resistant stainsteel. Dimensions L x H x W: 25 3/4  x 12 7/16  x 5 7/16 inch (654 x 316 x 138mm)Compartment Size l x h x w: 3 5/8 x 2 7/8 x 5 3/16 inch (92 x 73 x 131 mm)