Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for Microtiter Plates, 7 x 6 configuration, 42 Box Capacity
SDP
Catalog No. 503027958
Encompass_Preferred
For 96-Well and 384-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks allow for storage of 42 total 96/384-Well Microtiter Plates, Microtube Boxes or Micronic ROBO Racks. These freezer racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders for easy access and identification.  The drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.  Security lock devices are also available for each rack though ordered separately. Plate Capacity: 126 with lids 168 without lids7×6 Configuration imensions (DxWxH): 25 1/4 x 5 7/16 x 14 1/2 inches (642 x 138 x 369mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.
Catalog No. 50-302-7958 Supplier Alkali Scientific Supplier No. UFDMP76
For 96-Well and 384-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks allow for storage of 42 total 96/384-Well Microtiter Plates, Microtube Boxes or Micronic ROBO Racks. These freezer racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders for easy access and identification.  The drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.  Security lock devices are also available for each rack though ordered separately. Plate Capacity: 126 with lids 168 without lids7×6 Configuration imensions (DxWxH): 25 1/4 x 5 7/16 x 14 1/2 inches (642 x 138 x 369mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.