Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for Blood Sample Tubes – Fits 4ml, 4.5ml, 6ml, 7ml Tubes, 85 Capacity, 1 Drawer
SDP
Catalog No. 503027975
Encompass_Preferred
For Blood Sample Tubes4ml, 4.5ml, 6ml, 7ml Tube Capacity: 85 Tubes1 Drawer Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 85 tubes for 4ml, 4.5ml, 6ml, and 7ml sizes. This freezer rack features one drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 4 1/2 inches (424 x 140 x 114mm)
Catalog No. 50-302-7975 Supplier Alkali Scientific Supplier No. UFDRT061
For Blood Sample Tubes4ml, 4.5ml, 6ml, 7ml Tube Capacity: 85 Tubes1 Drawer Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 85 tubes for 4ml, 4.5ml, 6ml, and 7ml sizes. This freezer rack features one drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 4 1/2 inches (424 x 140 x 114mm)