Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 96 Deep-Well Microtiter Boxes; 6 x 7 Configuration, 42 Box Capacity
SDP
Catalog No. 503028048
Encompass_Preferred
For 96 Deep-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Micronic,Fluidx) H: 9/16″-1 3/4″ (14.4-44.9mm) Our Upright Freezer Drawer Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.  Racks are for 96 Deep-Well Microtiter Boxes.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Dimensions (DxWxH): 21 3/4 x 5 7/16 x 14 1/8 inches (552 x 138 x 358mm)Compartment Dimensions (l x h x w): 3 1/2 x 1 3/4 x 5 1/8 inch (89.4 x 45.4 x 130mm)
Catalog No. 50-302-8048 Supplier Alkali Scientific Supplier No. UFDDP67
For 96 Deep-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Micronic,Fluidx) H: 9/16″-1 3/4″ (14.4-44.9mm) Our Upright Freezer Drawer Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.  Racks are for 96 Deep-Well Microtiter Boxes.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Dimensions (DxWxH): 21 3/4 x 5 7/16 x 14 1/8 inches (552 x 138 x 358mm)Compartment Dimensions (l x h x w): 3 1/2 x 1 3/4 x 5 1/8 inch (89.4 x 45.4 x 130mm)