Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for Blood Sample Tubes – Fits 1mL Tubes, 240 Capacity, 2 Drawers
SDP
Catalog No. 503027821
Encompass_Preferred
For Blood Sample Tubes 1ml Tube Capacity: 240 Tubes2 Drawers Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 240 tubes for 1ml sizes. This freezer rack features two drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 6 1/16 inches (424 x 140 x 154mm)
Catalog No. 50-302-7821 Supplier Alkali Scientific Supplier No. UFDRT012
For Blood Sample Tubes 1ml Tube Capacity: 240 Tubes2 Drawers Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 240 tubes for 1ml sizes. This freezer rack features two drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 6 1/16 inches (424 x 140 x 154mm)