Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 96-well & 384-well Microtiter Plates, Holds 36 with Lids, 48 without Lids, 14 1/2 x 7 5/16 x 5 7/16"
SDP
Catalog No. 503027823
Encompass_Preferred
For 96-Well and 384-Well Microtiter Plates For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks allow for storage of 12 total 96/384-Well Microtiter Plates, Microtube Boxes or Micronic ROBO Racks. These freezer racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders for easy access and identification.  The drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up. Plate Capacity: 36 with lids 48 without lids 4×3 Configuration Dimensions (DxWxH): 14 5/8 x 5 7/16 x 7 5/16 inches (372 x 138 x 186mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.
Catalog No. 50-302-7823 Supplier Alkali Scientific Supplier No. UFDMP43
For 96-Well and 384-Well Microtiter Plates For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks allow for storage of 12 total 96/384-Well Microtiter Plates, Microtube Boxes or Micronic ROBO Racks. These freezer racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders for easy access and identification.  The drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up. Plate Capacity: 36 with lids 48 without lids 4×3 Configuration Dimensions (DxWxH): 14 5/8 x 5 7/16 x 7 5/16 inches (372 x 138 x 186mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.